5-Amino-6-methylpyridine-3-carbonitrile 97%

Lieferant: Apollo Scientific

Synonyme: 5-Amino-6-methylnicotinonitrile

OR480868-250MG
APOSOR480868-250MGEA 202 EUR
APOSOR480868-250MG
5-Amino-6-methylpyridine-3-carbonitrile 97%
5-Amino-6-methylpyridine-3-carbonitrile
Formel: C₇H₇N₃
Molecular Weight: 133,151 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD13175887
CAS-Nummer: 3308-01-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR