3,5-Diamino-2-hydroxybenzoic acid 95%
Lieferant: Apollo Scientific
OR480869-1G
APOSOR480869-1GEA
213
EUR
APOSOR480869-1G
3,5-Diamino-2-hydroxybenzoic acid 95%
3,5-Diamino-2-hydroxybenzoic acid
Formel:
C₇H₈N₂O₃ Molecular Weight: 168,15 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD09753462 CAS-Nummer: 112725-89-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...