tert-Butyl-2,4-dichloro-5,6-dihydropyrido[3,4-d]pyrimidine-7(8H)-carboxylate 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: tert-Butyl 2,4-dichloro-5,6-dihydropyrido[3,4-d]pyrimidine-7(8H)-carboxylate

OR480873-5G
APOSOR480873-5GEA 134 EUR
APOSOR480873-5G
tert-Butyl-2,4-dichloro-5,6-dihydropyrido[3,4-d]pyrimidine-7(8H)-carboxylate 97%
tert-Butyl-2,4-dichloro-5,6-dihydropyrido[3,4-d]pyrimidine-7(8H)-carboxylate
Formel: C₁₂H₁₅Cl₂N₃O₂
Molecular Weight: 304.1700 g/mol
Schmelzpunkt: 79…81 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD09025732
CAS-Nummer: 916420-27-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR