N1,N1-Dimethylpentane-1,5-diamine 95%

Lieferant: Apollo Scientific

Synonyme: 5-(Dimethylamino)amylamine

OR480879-1G
APOSOR480879-1GEA 237 EUR
APOSOR480879-1G
N1,N1-Dimethylpentane-1,5-diamine 95%
N1,N1-Dimethylpentane-1,5-diamine
Formel: C₇H₁₈N₂
Molecular Weight: 130,23 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00047978
CAS-Nummer: 3209-46-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR