3-Chloroimidazo[1,2-a]pyridine-7-carbohydrazide

Lieferant: Apollo Scientific
Achtung

OR48092-1G OR48092-250MG
APOSOR48092-1GEA 46.4 EUR
APOSOR48092-1G APOSOR48092-250MG
3-Chloroimidazo[1,2-a]pyridine-7-carbohydrazide
3-Chloroimidazo[1,2-a]pyridine-7-carbohydrazide
Formel: C₈H₇ClN₄O
Lagertemperatur: Raumtemperatur
MDL: MFCD32111678
CAS-Nummer: 2391987-06-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR