3-Chloroimidazo[1,2-a]pyridine-7-carboxylic acid
Lieferant: Apollo Scientific
|
Achtung
|
OR48093-1G
OR48093-250MG
APOSOR48093-1GEA
77
EUR
APOSOR48093-1G
APOSOR48093-250MG
3-Chloroimidazo[1,2-a]pyridine-7-carboxylic acid
3-Chloroimidazo[1,2-a]pyridine-7-carboxylic acid
Formel:
C₈H₅ClN₂O₂ Lagertemperatur: Raumtemperatur |
MDL:
MFCD21728893 CAS-Nummer: 1503889-91-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...