3-Chloroimidazo[1,2-a]pyridine-7-carboxylic acid

Lieferant: Apollo Scientific
Achtung

OR48093-1G OR48093-250MG
APOSOR48093-1GEA 77 EUR
APOSOR48093-1G APOSOR48093-250MG
3-Chloroimidazo[1,2-a]pyridine-7-carboxylic acid
3-Chloroimidazo[1,2-a]pyridine-7-carboxylic acid
Formel: C₈H₅ClN₂O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD21728893
CAS-Nummer: 1503889-91-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR