3,5-Dichloroimidazo[1,2-a]pyridine-2-carboxylic acid

Lieferant: Apollo Scientific
Achtung

OR48094-250MG
APOSOR48094-250MGEA 169 EUR
APOSOR48094-250MG
3,5-Dichloroimidazo[1,2-a]pyridine-2-carboxylic acid
3,5-Dichloroimidazo[1,2-a]pyridine-2-carboxylic acid
Formel: C₈H₄Cl₂N₂O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD11846271
CAS-Nummer: 1227954-34-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR