1,6-Anhydro-β-D-glucose
Lieferant: Apollo Scientific
|
Achtung
|
OR4810T-100G
OR4810T-25G
OR4810T-5G
APOSOR4810T-100GEA
741
EUR
APOSOR4810T-100G
APOSOR4810T-25G
APOSOR4810T-5G
1,6-Anhydro-β-D-glucose
1,6-Anhydro-beta-D-glucose
Formel:
C₆H₁₀O₅ Schmelzpunkt: 182…184 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00063248 CAS-Nummer: 498-07-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...