Ethyl 2-(3-aminooxetan-3-yl)acetate hemioxalate

Lieferant: Apollo Scientific

OR48129-1G
APOSOR48129-1GEA 589 EUR
APOSOR48129-1G
Ethyl 2-(3-aminooxetan-3-yl)acetate hemioxalate
Ethyl 2-(3-aminooxetan-3-yl)acetate hemioxalate
Formel: 2C₇H₁₃NO₃·C₂H₂O₄
Molecular Weight: 408,401 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD27986902
CAS-Nummer: 1523618-27-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR