3,5-Bis(benzyloxy)benzoesäure 96%
Lieferant: Apollo Scientific
Synonyme:
3,5-Bis(benzyloxy)benzoic acid
OR4816-25G
APOSOR4816-25GEA
43.5
EUR
APOSOR4816-25G
3,5-Bis(benzyloxy)benzoesäure 96%
3,5-Bis(benzyloxy)benzoesäure
Formel:
C₂₁H₁₈O₄ Molecular Weight: 334,37 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00191341 CAS-Nummer: 28917-43-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...