3-Chloro-6-(1-methylhydrazino)pyridazine

Lieferant: Apollo Scientific
Achtung

OR4821-1G
APOSOR4821-1GEA 342 EUR
APOSOR4821-1G
3-Chloro-6-(1-methylhydrazino)pyridazine
3-Chloro-6-(1-methylhydrazino)pyridazine
Formel: C₅H₇ClN₄
Lagertemperatur: Kühlschrank
MDL: MFCD00052661
CAS-Nummer: 76953-33-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR