2'-Brom-4',5'-(ethylendioxy)acetophenon 98%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
1-(7-Bromo-2,3-dihydrobenzo[b][1,4]dioxin-6-yl)ethanone
OR4822-1G
OR4822-250MG
OR4822-5G
APOSOR4822-1GEA
161
EUR
APOSOR4822-1G
APOSOR4822-250MG
APOSOR4822-5G
2'-Brom-4',5'-(ethylendioxy)acetophenon 98%
2'-Brom-4',5'-(ethylendioxy)acetophenon
Formel:
C₁₀H₉BrO₃ Schmelzpunkt: 85…87 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00168681 CAS-Nummer: 59820-90-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...