5-Bromo-2-ethylpyrimidine-4-carboxylic acid 97%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
5-Bromo-4-carboxy-2-ethyl-1,3-diazine, 5-Bromo-4-carboxy-2-ethylpyrimidine
OR48242-250MG
APOSOR48242-250MGEA
165
EUR
APOSOR48242-250MG
5-Bromo-2-ethylpyrimidine-4-carboxylic acid 97%
5-Bromo-2-ethylpyrimidine-4-carboxylic acid
Formel:
C₇H₇BrN₂O₂ Molecular Weight: 231,047 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD11870588 CAS-Nummer: 1235450-86-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...