(S)-(-)-4-Brom-α-methylbenzylalkohol 98%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
(S)-1-(4-Bromophenyl)ethanol
OR48261-10G
OR48261-1G
OR48261-5G
APOSOR48261-10GEA
345
EUR
APOSOR48261-10G
APOSOR48261-1G
APOSOR48261-5G
(S)-(-)-4-Brom-α-methylbenzylalkohol 98%
(S)-(-)-4-Brom-α-methylbenzylalkohol
Formel:
C₈H₉BrO Molecular Weight: 201.06 g/mol Dichte: 1.322 g/cm³ (25 °C) Flammpunkt: >110 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD06201860 CAS-Nummer: 100760-04-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...