Ethyl 4-[(3-chloro-4-methoxybenzyl)amino]-2-(methylthio)pyrimidine-5-carboxylate

Lieferant: Apollo Scientific
Achtung

Synonyme: Ethyl 4-((3-chloro-4-methoxybenzyl)amino)-2-(methylthio)pyrimidine-5-carboxylate, 4-[(3-Chloro-4-methoxybenzyl)amino]-5-(ethoxycarbonyl)-2-(methylthio)pyrimidine, Ethyl 4-[(3-chloro-4-methoxybenzyl)amino]-2-(methylsulphanyl)pyrimidine-5-carboxylate

OR48263-1G OR48263-25G OR48263-5G
APOSOR48263-1GEA 21.9 EUR
APOSOR48263-1G APOSOR48263-25G APOSOR48263-5G
Ethyl 4-[(3-chloro-4-methoxybenzyl)amino]-2-(methylthio)pyrimidine-5-carboxylate
Ethyl 4-[(3-chloro-4-methoxybenzyl)amino]-2-(methylthio)pyrimidine-5-carboxylate
Formel: C₁₆H₁₈ClN₃O₃S
Lagertemperatur: Kühlschrank
MDL: MFCD17012695
CAS-Nummer: 330785-81-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR