(2R)-2-(4-Hydroxyphenyl)propanoic acid 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: (R)-(4-Hydroxyphenyl)(methyl)acetic acid, (2R)-2-(4-Hydroxyphenyl)propionic acid, 4-[(1R)-1-Carboxyethyl]phenol

OR48264-1G OR48264-250MG OR48264-5G
APOSOR48264-1GEA 406 EUR
APOSOR48264-1G APOSOR48264-250MG APOSOR48264-5G
(2R)-2-(4-Hydroxyphenyl)propanoic acid 98%
(2R)-2-(4-Hydroxyphenyl)propanoic acid
Formel: C₉H₁₀O₃
Schmelzpunkt: 156…158 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD21606316
CAS-Nummer: 59092-88-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR