(3-exo)-3-(3-Isopropyl-5-methyl-4H-1,2,4-triazol-4-yl]-8-azabicyclo[3.2.1]octane 95+%

Lieferant: Apollo Scientific

Synonyme: 4-[(3-exo)-8-Azabicyclo[3.2.1]oct-3-yl]-3-isopropyl-5-methyl-4H-1,2,4-triazole, (3-exo)-3-[3-Methyl-5-(prop-2-yl)-4H-1,2,4-triazol-4-yl]-8-azabicyclo[3.2.1]octane, exo-3-(3-Isopropyl-5-methyl-4H-1,2,4-triazol-4-yl)-8-azabicyclo[3.2.1]octane

OR48267-250MG
APOSOR48267-250MGEA 26.1 EUR
APOSOR48267-250MG
(3-exo)-3-(3-Isopropyl-5-methyl-4H-1,2,4-triazol-4-yl]-8-azabicyclo[3.2.1]octane 95+%
(3-exo)-3-(3-Isopropyl-5-methyl-4H-1,2,4-triazol-4-yl]-8-azabicyclo[3.2.1]octane
Formel: C₁₃H₂₂N₄
Molecular Weight: 234,341 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD12912695
CAS-Nummer: 423165-07-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR