2-Amino-N-(2-chlor-6-methylphenyl)-1,3-thiazol-5-carboxamid 98%
Lieferant: Apollo Scientific
OR48268-25G
OR48268-5G
OR48268-100G
APOSOR48268-25GEA
88.6
EUR
APOSOR48268-25G
APOSOR48268-5G
APOSOR48268-100G
2-Amino-N-(2-chlor-6-methylphenyl)-1,3-thiazol-5-carboxamid 98%
2-Amino-N-(2-chloro-6-methylphenyl)thiazole-5-carboxamide
Formel:
C₁₁H₁₀ClN₃OS Lagertemperatur: Kühlschrank |
MDL:
MFCD10000630 CAS-Nummer: 302964-24-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...