2-Amino-N-(2-chlor-6-methylphenyl)-1,3-thiazol-5-carboxamid 98%

Lieferant: Apollo Scientific

OR48268-25G
APOSOR48268-25GEA 88.6 EUR
APOSOR48268-25G
2-Amino-N-(2-chlor-6-methylphenyl)-1,3-thiazol-5-carboxamid 98%
2-Amino-N-(2-chloro-6-methylphenyl)thiazole-5-carboxamide
Formel: C₁₁H₁₀ClN₃OS
Lagertemperatur: Kühlschrank
MDL: MFCD10000630
CAS-Nummer: 302964-24-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR