4-Bromo-2-chloro-5-nitrophenol 95%

Lieferant: Apollo Scientific
Achtung

OR48270-1G OR48270-250MG OR48270-5G
APOSOR48270-1GEA 574 EUR
APOSOR48270-1G APOSOR48270-250MG APOSOR48270-5G
4-Bromo-2-chloro-5-nitrophenol 95%
4-Bromo-2-chloro-5-nitrophenol
Formel: C₆H₃BrClNO₃
Lagertemperatur: Raumtemperatur
MDL: MFCD31700576
CAS-Nummer: 2091684-77-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR