5-(2-Chlorphenyl)oxazol 95%
Lieferant: Apollo Scientific
|
Achtung
|
OR4832-1G
OR4832-250MG
APOSOR4832-1GEA
139
EUR
APOSOR4832-1G
APOSOR4832-250MG
5-(2-Chlorphenyl)oxazol 95%
5-(2-Chlorophenyl)oxazole
Formel:
C₉H₆ClNO Schmelzpunkt: 33…35 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00084973 CAS-Nummer: 89808-74-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...