2H-1,4-Benzoxazine-3(4H)-thione

Lieferant: Apollo Scientific

Synonyme: 3,4-Dihydro-3-thioxo-2H-1,4-benzoxazine

OR4833-1G
APOSOR4833-1GEA 205 EUR
APOSOR4833-1G
2H-1,4-Benzoxazine-3(4H)-thione
2H-1,4-Benzoxazine-3(4H)-thione
Formel: C₈H₇NOS
Molecular Weight: 165,212 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00269330
CAS-Nummer: 14183-51-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR