Ethyl-6-chlor-2-methylimidazo[1,2-a]pyridin-3-carboxylat

Lieferant: Apollo Scientific
Achtung

OR4875-1G
APOSOR4875-1GEA 644 EUR
APOSOR4875-1G
Ethyl-6-chlor-2-methylimidazo[1,2-a]pyridin-3-carboxylat
Ethyl-6-chlor-2-methylimidazo[1,2-a]pyridin-3-carboxylat
Formel: C₁₁H₁₁ClN₂O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD00269623
CAS-Nummer: 330858-13-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR