6-Chloro-2-phenylimidazo[1,2-a]pyridine-3-carboxylic acid

Lieferant: Apollo Scientific
Achtung

OR4876-100MG
APOSOR4876-100MGEA 109 EUR
APOSOR4876-100MG
6-Chloro-2-phenylimidazo[1,2-a]pyridine-3-carboxylic acid
6-Chloro-2-phenylimidazo[1,2-a]pyridine-3-carboxylic acid
Formel: C₁₄H₉ClN₂O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD00665805
CAS-Nummer: 220465-51-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR