Ethyl-6-chlor-2-phenylimidazo[1,2-a]pyridin-3-carboxylat
Lieferant: Apollo Scientific
|
Achtung
|
OR4877-250MG
APOSOR4877-250MGEA
120
EUR
APOSOR4877-250MG
Ethyl-6-chlor-2-phenylimidazo[1,2-a]pyridin-3-carboxylat
Ethyl-6-chlor-2-phenylimidazo[1,2-a]pyridin-3-carboxylat
Formel:
C₁₆H₁₃ClN₂O₂ Schmelzpunkt: 124…125 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00139524 CAS-Nummer: 193979-37-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...