3-(2-Acetamidoethyl)-1H-indol-5-yl acetate

Lieferant: Apollo Scientific
Achtung

Synonyme: 3-[2-(Acetylamino)ethyl]-1H-indol-5-yl acetate, 5-Acetoxy-N-acetyltryptamine

OR4891-100MG
APOSOR4891-100MGEA 386 EUR
APOSOR4891-100MG
3-(2-Acetamidoethyl)-1H-indol-5-yl acetate
3-(2-Acetamidoethyl)-1H-indol-5-yl acetate
Formel: C₁₄H₁₆N₂O₃
Molecular Weight: 260.288 g/mol
Schmelzpunkt: 121 °C
Lagertemperatur: Gefrierschrank
MDL: MFCD00209948
CAS-Nummer: 28026-16-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR