4-Morpholinoanilin 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: 4-(4-Morpholinyl)aniline

OR4896-100G OR4896-25G OR4896-5G
APOSOR4896-100GEA 115 EUR
APOSOR4896-100G APOSOR4896-25G APOSOR4896-5G
4-Morpholinoanilin 98%
4-Morpholinoanilin
Formel: C₁₀H₁₄N₂O
Molecular Weight: 178.23 g/mol
Schmelzpunkt: 132…135 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00006169
CAS-Nummer: 2524-67-6
EINECS: 219-760-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR