3,4-Dichlorthiobenzamid

Lieferant: Apollo Scientific

Synonyme: 3,4-Dichlorthiobenzoesäureamid

OR49004-10G OR49004-1G OR49004-5G
APOSOR49004-10GEA 522 EUR
APOSOR49004-10G APOSOR49004-1G APOSOR49004-5G
3,4-Dichlorthiobenzamid
3,4-Dichlorthiobenzamid
Formel: C₇H₅Cl₂NS
Schmelzpunkt: 133…137 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD04973321
CAS-Nummer: 22179-73-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR