1-(Chloroacetyl)piperidine
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
2-Chloro-1-(piperidin-1-yl)ethan-1-one, 1-(2-Chloroethanoyl)piperidine
OR49021-1G
APOSOR49021-1GEA
109
EUR
APOSOR49021-1G
1-(Chloroacetyl)piperidine
1-(Chloroacetyl)piperidine
Formel:
C₇H₁₂ClNO Molecular Weight: 161,629 g/mol Siedepunkt: 83…85 °C (0,075 mm Hg) Lagertemperatur: Raumtemperatur |
MDL:
MFCD01095825 CAS-Nummer: 1440-60-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...