Imidazo[1,5-a]pyrazin-8-amine
Lieferant: Apollo Scientific
OR49048-250MG
APOSOR49048-250MGEA
232
EUR
APOSOR49048-250MG
Imidazo[1,5-a]pyrazin-8-amine
Imidazo[1,5-a]pyrazin-8-amine
Formel:
C₆H₆N₄ Molecular Weight: 134,139 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD09835041 CAS-Nummer: 26538-77-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...