4-(1H-Pyrazol-4-yl)-7H-pyrrolo[2,3-d]pyrimidine
Lieferant: Apollo Scientific
OR49052-1G
APOSOR49052-1GEA
567
EUR
APOSOR49052-1G
4-(1H-Pyrazol-4-yl)-7H-pyrrolo[2,3-d]pyrimidine
4-(1H-Pyrazol-4-yl)-7H-pyrrolo[2,3-d]pyrimidine
Formel:
C₉H₇N₅ Molecular Weight: 185.185 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD20683197 CAS-Nummer: 952518-97-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...