7-Methoxyquinazoline

Lieferant: Apollo Scientific

OR49055-250MG
APOSOR49055-250MGEA 835 EUR
APOSOR49055-250MG
7-Methoxyquinazoline
7-Methoxyquinazoline
Formel: C₉H₈N₂O
Molecular Weight: 160,173 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD18378811
CAS-Nummer: 10105-37-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR