(1S,3R)-Cyclopentane-1,3-diol

Lieferant: Apollo Scientific

Synonyme: cis-Cyclopentane-1,3-diol

OR49056-1G OR49056-250MG
APOSOR49056-1GEA 922 EUR
APOSOR49056-1G APOSOR49056-250MG
(1S,3R)-Cyclopentane-1,3-diol
(1S,3R)-Cyclopentane-1,3-diol
Formel: C₅H₁₀O₂
Molecular Weight: 102.132 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD13619671
CAS-Nummer: 16326-97-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR