Ethyl 5-hydroxypyrazolo[1,5-a]pyrimidine-3-carboxylate
Lieferant: Apollo Scientific
OR49062-1G
APOSOR49062-1GEA
342
EUR
APOSOR49062-1G
Ethyl 5-hydroxypyrazolo[1,5-a]pyrimidine-3-carboxylate
Ethyl 5-hydroxypyrazolo[1,5-a]pyrimidine-3-carboxylate
Formel:
C₉H₉N₃O₃ Molecular Weight: 207.186 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD21607426 CAS-Nummer: 1026456-01-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...