Di(1-adamantyl)-n-butylphosphine 98%
Lieferant: Apollo Scientific
|
Achtung
|
OR49071-1G
OR49071-25G
OR49071-5G
APOSOR49071-1GEA
49.3
EUR
APOSOR49071-1G
APOSOR49071-25G
APOSOR49071-5G
Di(1-adamantyl)-n-butylphosphine 98%
Di(1-adamantyl)-n-butylphosphine
Formel:
C₂₄H₃₉P Lagertemperatur: Raumtemperatur |
MDL:
MFCD05861606 CAS-Nummer: 321921-71-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...