2-(Chloromethyl)-4-hydroxyquinazoline

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-(Chloromethyl)quinazlin-4-ol

OR4920-1G
APOSOR4920-1GEA 39.3 EUR
APOSOR4920-1G
2-(Chloromethyl)-4-hydroxyquinazoline
2-(Chloromethyl)-4-hydroxyquinazoline
Formel: C₉H₇ClN₂O
Molecular Weight: 194,618 g/mol
Schmelzpunkt: 249…253 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00665825
CAS-Nummer: 3817-05-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR