4-(4-Chlorophenyl)pyrimidin-2-amine

Lieferant: Apollo Scientific
Achtung

OR4928-1G
APOSOR4928-1GEA 203 EUR
APOSOR4928-1G
4-(4-Chlorophenyl)pyrimidin-2-amine
4-(4-Chlorophenyl)pyrimidin-2-amine
Formel: C₁₀H₈ClN₃
Schmelzpunkt: 170…174 °C
Lagertemperatur: Kühlschrank
MDL: MFCD00665913
CAS-Nummer: 133256-51-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR