4,4'-Biphenol 99%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
Biphenyl-4,4'-diol, 4,4'-Dihydroxybiphenyl, 1-Hydroxy-4-(4-hydroxyphenyl)benzol
OR4930-2.5KG
APOSOR4930-2.5KGEA
162
EUR
APOSOR4930-2.5KG
4,4'-Biphenol 99%
4,4'-Biphenol
Formel:
C₁₂H₁₀O₂ Molecular Weight: 186,21 g/mol Schmelzpunkt: 280…282 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00002348 CAS-Nummer: 92-88-6 UN: 3077 ADR: 9+EHS,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...