2,1,3-Benzothiadiazol-4-sulfonylchlorid
Lieferant: Apollo Scientific
|
Gefahr
|
Synonyme:
Benzo[c][1,2,5]thiadiazole-4-sulphonyl chloride
OR4940-250MG
APOSOR4940-250MGEA
21.9
EUR
APOSOR4940-250MG
2,1,3-Benzothiadiazol-4-sulfonylchlorid
2,1,3-Benzothiadiazol-4-sulfonylchlorid
Formel:
C₆H₃ClN₂O₂S₂ Molecular Weight: 234,68 g/mol Schmelzpunkt: 148…150 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00068049 CAS-Nummer: 73713-79-8 UN: 1759 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...