2',3',4'-Trihydroxyacetophenon 97%
Lieferant: Apollo Scientific
|
Achtung
|
OR4951-100G
OR4951-25G
APOSOR4951-100GEA
173
EUR
APOSOR4951-100G
APOSOR4951-25G
2',3',4'-Trihydroxyacetophenon 97%
2',3',4'-Trihydroxyacetophenon
Formel:
C₈H₈O₄ Schmelzpunkt: 169…172 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00002193 CAS-Nummer: 528-21-2 EINECS: 208-430-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...