5-Iodvanillin 97%

Lieferant: Apollo Scientific

Synonyme: 4-Hydroxy-3-iodo-5-methoxybenzaldehyde

OR4972-100G
APOSOR4972-100GEA 241 EUR
APOSOR4972-100G
5-Iodvanillin 97%
5-Iodvanillin
Formel: C₈H₇IO₃
Molecular Weight: 278,05 g/mol
Schmelzpunkt: 180…182 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00006941
CAS-Nummer: 5438-36-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR