3,5-Dimethoxybenzoesäure 97%
Lieferant: Apollo Scientific
|
Achtung
|
OR4991-500G
APOSOR4991-500GEA
224
EUR
APOSOR4991-500G
3,5-Dimethoxybenzoesäure 97%
3,5-Dimethoxybenzoic acid
Formel:
C₉H₁₀O₄ Molecular Weight: 182.18 g/mol Siedepunkt: 341 Schmelzpunkt: 181…183 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00002502 CAS-Nummer: 1132-21-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...