1-Acetyl-4-cyclopropylaminopiperidine 95+%

Lieferant: Apollo Scientific
Achtung

Synonyme: 1-[4-(Cyclopropylamino)piperidin-1-yl]ethan-1-one

OR4996-1G
APOSOR4996-1GEA 34.8 EUR
APOSOR4996-1G
1-Acetyl-4-cyclopropylaminopiperidine 95+%
1-Acetyl-4-cyclopropylaminopiperidine
Formel: C₁₀H₁₈N₂O
Molecular Weight: 182,27 g/mol
Siedepunkt: 120 °C/0,075mmHg
Lagertemperatur: Raumtemperatur
MDL: MFCD01941331
CAS-Nummer: 387358-46-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR