5-(3,4-Dichlorphenyl)isoxazol

Lieferant: Apollo Scientific
Achtung

OR5-250MG
APOSOR5-250MGEA 58 EUR
APOSOR5-250MG
5-(3,4-Dichlorphenyl)isoxazol
5-(3,4-Dichlorophenyl)isoxazole
Formel: C₉H₅Cl₂NO
Lagertemperatur: Raumtemperatur
CAS-Nummer: 138716-63-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR