2-Amino-3-chloro-5-methylphenol 97%
Lieferant: Apollo Scientific
|
Achtung
|
OR500003-250MG
APOSOR500003-250MGEA
104
EUR
APOSOR500003-250MG
2-Amino-3-chloro-5-methylphenol 97%
2-Amino-3-chloro-5-methylphenol
Formel:
C₇H₈ClNO Molecular Weight: 157,598 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD28551485 CAS-Nummer: 1780449-40-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...