2,4-Dichlor-6-methoxyanilin 95+%

Lieferant: Apollo Scientific
Achtung

OR500006-250MG
APOSOR500006-250MGEA 55.1 EUR
APOSOR500006-250MG
2,4-Dichlor-6-methoxyanilin 95+%
2,4-Dichlor-6-methoxyanilin
Formel: C₇H₇Cl₂NO
Molecular Weight: 192,04 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD20693704
CAS-Nummer: 93839-14-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR