4-Amino-2,5-dibromophenol 95+%

Lieferant: Apollo Scientific
Achtung

OR500009-250MG
APOSOR500009-250MGEA 133 EUR
APOSOR500009-250MG
4-Amino-2,5-dibromophenol 95+%
4-Amino-2,5-dibromophenol
Formel: C₆H₅Br₂NO
Molecular Weight: 266,918 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD28040627
CAS-Nummer: 872272-33-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR