Methyl 4,6-dibromopicolinate 95+%

Lieferant: Apollo Scientific
Achtung

OR500014-250MG
APOSOR500014-250MGEA 66.7 EUR
APOSOR500014-250MG
Methyl 4,6-dibromopicolinate 95+%
Methyl 4,6-dibromopicolinate
Formel: C₇H₅Br₂NO₂
Lagertemperatur: Kühlschrank
MDL: MFCD14582014
CAS-Nummer: 1206248-47-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR