4-Bromo-5-chloro-2-methylpyridine 95+%
Lieferant: Apollo Scientific
|
Achtung
|
OR500015-100MG
OR500015-250MG
APOSOR500015-100MGEA
155
EUR
APOSOR500015-100MG
APOSOR500015-250MG
4-Bromo-5-chloro-2-methylpyridine 95+%
4-Bromo-5-chloro-2-methylpyridine
Formel:
C₆H₅BrClN Molecular Weight: 206.468 g/mol Lagertemperatur: Kühlschrank |
MDL:
MFCD17014484 CAS-Nummer: 1211529-34-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...